2-Bromo-4-fluorobenzoic acid ≥98%
Supplier: BeanTown Chemical
134555-1G
134555-25G
134555-5G
BT134555-1GEA
24
USD
BT134555-1G
BT134555-25G
BT134555-5G
2-Bromo-4-fluorobenzoic acid ≥98%
2-Bromo-4-fluorobenzoic acid
CAS: 1006-41-3; MDL No: MFCD00055370
Powder; Molecular Formula: C7H4BrFO2; MW: 219.01
Melting Point: 172-176°
- Powder
Formula:
C₇H₄BrFO₂ Melting Pt: 172…176 °C |
MDL Number:
MFCD00055370 CAS Number: 1006-41-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...