Indium(III) acetylacetonate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Indium (III) tris(2,4-pentanedionato-O,O'), Indium (III) 2,4-pentanedionate, Tris(2,4-pentanedionato-O,O')indium (III)
134540-1G
134540-25G
134540-5G
BT134540-1GEA
26
USD
BT134540-1G
BT134540-25G
BT134540-5G
Indium(III) acetylacetonate ≥98%
Indium(III) acetylacetonate
CAS: 14405-45-9; EC No: 238-378-6; MDL No: MFCD00013494
Powder; Molecular Formula: C15H21InO6; MW: 412.15
Melting Point: 187-189°
- Powder
Formula:
C₁₅H₂₁InO₆ MW: 412.15 g/mol Melting Pt: 187…189 °C |
MDL Number:
MFCD00013494 CAS Number: 14405-45-9 EINECS: 238-378-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...