Boc-Pro-OH ≥99%

Supplier: BeanTown Chemical

Synonyms: N-(tert-Butoxycarbonyl)-L-proline, N-Boc-L-proline, N-α-t-Boc-L-Proline, BOC-L-Proline, (S)-1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid

134535-25G 134535-5G
BT134535-25GEA 81.43 USD
BT134535-25G BT134535-5G
Boc-Pro-OH ≥99%
Boc-Pro-OH

CAS: 15761-39-4; EC No: 239-848-3; MDL No: MFCD00037324 Crystalline; Molecular Formula: C10H17NO4; MW: 215.25 Melting Point: 133-135° Optical Rotation: [α]20/D -60°, c = 1 in acetic acid


  • Optical rotation: [α]20/D -60°, c = 1 in acetic acid
  • Crystalline

Formula: C₁₀H₁₇NO₄
Melting Pt: 133…135 °C
MDL Number: MFCD00037324
CAS Number: 15761-39-4
EINECS: 239-848-3

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR