1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene ≥99%
Supplier: BeanTown Chemical
Synonyms:
1-Ethyl-4-(p-tolylethynyl)benzene, 1-Ethyl-4-[(p-tolyl)ethynyl]benzene, 1-Ethyl-4-[(4-methylphenyl)ethynyl]benzene, 1-(4-Ethylphenyl)-2-(p-tolyl)acetylene
134530-1G
134530-5G
BT134530-1GEA
58
USD
BT134530-1G
BT134530-5G
1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene ≥99%
1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene
CAS: 22692-80-4; MDL No: MFCD04038793
Crystalline/Powder; Molecular Formula: C17H16; MW: 220.31
Melting Point: 71-74°; Boiling Point: 135-136°/1 mmHg
- Crystalline/Powder
Formula:
C₁₇H₁₆ MW: 220.315 g/mol Boiling Pt: 135…136 °C/1 mmHg Melting Pt: 71…74 °C |
MDL Number:
MFCD04038793 CAS Number: 22692-80-4 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...