Thieno[3,2-b]pyridin-7-ol ≥96%

Supplier: BeanTown Chemical

Synonyms: 7-Hydroxythieno[3,2-b]pyridine

134515-1G 134515-5G
BT134515-1GEA 52 USD
BT134515-1G BT134515-5G
Thieno[3,2-b]pyridin-7-ol ≥96%
Thieno[3,2-b]pyridin-7-ol

CAS: 107818-20-2; MDL No: MFCD01863636 Solid; Molecular Formula: C7H5NOS; MW: 151.19 Melting Point: 230-235°


  • Solid

Formula: C₇H₅NOS
MW: 151.183 g/mol
Melting Pt: 230…235 °C
MDL Number: MFCD01863636
CAS Number: 107818-20-2

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR