Gold tin eutectic alloy (80:20 wt%) ≥99.99% (trace metals basis), slug, Length 6.35 mm, Diameter 6.35 mm

Supplier: BeanTown Chemical

134495-1PC 134495-5PC
BT134495-1PCEA 392 USD
BT134495-1PC BT134495-5PC
Gold tin eutectic alloy (80:20 wt%) ≥99.99% (trace metals basis), slug, Length 6.35 mm, Diameter 6.35 mm
Gold tin eutectic alloy (80:20 wt%)

MDL No: MFCD02091709 Solid; Formula: Au:Sn; 80:20 wt%


  • Solid

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR