Tris(4-methoxyphenyl)phosphine
Supplier: BeanTown Chemical
Synonyms:
Tris(p-methoxyphenyl)phosphine
134475-1G
134475-25G
134475-5G
BT134475-1GEA
35
USD
BT134475-1G
BT134475-25G
BT134475-5G
Tris(4-methoxyphenyl)phosphine
Tris(4-methoxyphenyl)phosphine
CAS: 855-38-9; EC No: 212-723-0; MDL No: MFCD00014896
Powder; Linear Formula: (CH3OC6H4)3P; MW: 352.36
Melting Point: 131-134°
Air Sensitive
- Air Sensitive
- Powder
Formula:
C₂₁H₂₁O₃P MW: 352.37 g/mol Melting Pt: 131…134 °C |
MDL Number:
MFCD00014896 CAS Number: 855-38-9 EINECS: 212-723-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...