6-Fluoroanthranilic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Amino-6-fluorobenzoic acid
134460-1G
134460-25G
134460-5G
BT134460-1GEA
38
USD
BT134460-1G
BT134460-25G
BT134460-5G
6-Fluoroanthranilic acid ≥98%
6-Fluoroanthranilic acid
CAS: 434-76-4; MDL No: MFCD00067781
Solid; Molecular Formula: C7H6FNO2 ; MW: 155.13
Melting Point: 167-169°
- Solid
Formula:
C₇H₆FNO₂ MW: 155.13 g/mol Melting Pt: 167…169 °C |
MDL Number:
MFCD00067781 CAS Number: 434-76-4 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...