1,4-Benzoquinone dioxime ≥95%
Supplier: BeanTown Chemical
Synonyms:
p-Quinone Dioxime
134445-1KG
134445-250G
134445-50G
BT134445-1KGEA
500
USD
BT134445-1KG
BT134445-250G
BT134445-50G
1,4-Benzoquinone dioxime ≥95%
1,4-Benzoquinone dioxime
CAS: 105-11-3; EC No: 203-271-5; MDL No: MFCD00063636; RTECS: DK4900000
Crystalline/Powder; Molecular Formula: C6H6N2O2; MW: 138.12
Melting Point: 243° (decomposes)
- RTECS: DK4900000
- Crystalline/Powder
Formula:
C₆H₆N₂O₂ MW: 138.13 g/mol Melting Pt: 243 °C (dec.) |
MDL Number:
MFCD00063636 CAS Number: 105-11-3 EINECS: 203-271-5 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...