2-(4-Chloro-3-nitrobenzoyl)benzoic acid ≥98%

Supplier: BeanTown Chemical

134370-250G 134370-50G
BT134370-250GEA 207 USD
BT134370-250G BT134370-50G
2-(4-Chloro-3-nitrobenzoyl)benzoic acid ≥98%
2-(4-Chloro-3-nitrobenzoyl)benzoic acid

CAS: 85-54-1; EC No: 201-613-8; MDL No: MFCD00007082 Powder; Molecular Formula: C14H7ClNO5; MW: 305.67 Melting Point: 198-201°


  • Powder

Formula: C₁₄H₈ClNO₅
MW: 305.67 g/mol
Melting Pt: 198…201 °C
MDL Number: MFCD00007082
CAS Number: 85-54-1
EINECS: 201-613-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR