Cesium metasilicate ≥99%
Supplier: BeanTown Chemical
134355-100G
134355-25G
BT134355-100GEA
426
USD
BT134355-100G
BT134355-25G
Cesium metasilicate ≥99%
Cesium metasilicate
CAS: 15586-77-3; MDL No: MFCD00798519
Powder; Linear Formula: Cs2SiO3; MW: 341.89
- Powder
Formula:
Cs₂O₃Si MW: 341.893 g/mol |
MDL Number:
MFCD00798519 CAS Number: 15586-77-3 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...