2-Fluoro-3-nitrobenzoic acid ≥95%

Supplier: BeanTown Chemical

134345-1G 134345-5G
BT134345-1GEA 36 USD
BT134345-1G BT134345-5G
2-Fluoro-3-nitrobenzoic acid ≥95%
2-Fluoro-3-nitrobenzoic acid

CAS: 317-46-4; MDL No: MFCD06797825 Solid; Molecular Formula: C7H4FNO4; MW: 185.11 Air Sensitive


  • Air Sensitive
  • Solid

Formula: C₇H₄FNO₄
MDL Number: MFCD06797825
CAS Number: 317-46-4

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR