Zinc stearate

Supplier: BeanTown Chemical

Synonyms: Stearic acid zinc salt, Zinc(II) stearate

134300-1KG 134300-250G 134300-50G
BT134300-1KGEA 50.69 USD
BT134300-1KG BT134300-250G BT134300-50G
Zinc stearate
Zinc stearate

CAS: 557-05-1; EC No: 209-151-9; MDL No: MFCD00013031; RTECS: WI4390000 Powder; Linear Formula: Zn[CH3(CH2)16CO2]2 ; Molecular Formula: C36H70O4Zn; MW: 632.33 Melting Point: 128-130°; Boiling Point: 135° Density (g/mL): 0.403


  • RTECS: WI4390000
  • Powder

Formula: C₃₆H₇₀O₄Zn
MW: 632.34 g/mol
Boiling Pt: 135 °C
Melting Pt: 128…130 °C
Density: 0.403 g/mL
MDL Number: MFCD00013031
CAS Number: 557-05-1
EINECS: 209-151-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8ad7f3e101681798edbd54e57ad5d2ac82b2e5d&vid=435308cdcb35862fe6cc06384da2ef10d84c5700&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR