1,1,1-Tris(hydroxymethyl)propane ≥98%

Supplier: BeanTown Chemical

Synonyms: 1,1,1-Trimethylolpropane, Propylidynetrimethanol, Trimethylol propane, Propylidynetris(methanol), 2-Ethyl-2-(hydroxymethyl)-1,3-propanediol

134225-10KG 134225-2.5KG 134225-500G
BT134225-10KGEA 282 USD
BT134225-10KG BT134225-2.5KG BT134225-500G
1,1,1-Tris(hydroxymethyl)propane ≥98%
1,1,1-Tris(hydroxymethyl)propane

CAS: 77-99-6; EC No: 201-074-9; MDL No: MFCD00004694; RTECS: TY6470000 Crystalline; Linear Formula: CH3CH2C(CH2OH)3; Molecular Formula: C6H14O3; MW: 134.17 Melting Point: 56-58°; Boiling Point: 159-161°/2 mmHg; Flash point: 172°C (342°F)


  • RTECS: TY6470000
  • Crystalline

Formula: C₆H₁₄O₃
MW: 134.18 g/mol
Boiling Pt: 159…161 °C/2 mmHg
Melting Pt: 56…58 °C
Flash Pt: 172 °C (342 °F)
MDL Number: MFCD00004694
CAS Number: 77-99-6
EINECS: 201-074-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR