Erioglaucine disodium salt 60% (dye content)
Supplier: BeanTown Chemical
Synonyms:
FD and C Blue No. 1, Brilliant Blue FCF, Erioglaucin A, Sodium 2-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)benzenesulphonate, Alphazurine FG, Erioglaucine sodium salt, CI 42090, Acid blue 9, Food Blue No.1
134220-25G
134220-5G
BT134220-25GEA
45.69
USD
BT134220-25G
BT134220-5G
Erioglaucine disodium salt 60% (dye content)
Erioglaucine disodium salt
CAS: 3844-45-9; EC No: 223-339-8; MDL No: MFCD00001657; RTECS: BQ4725000
Powder; Molecular Formula: C37H34Na2N2O9S3; MW: 792.85
Melting Point: 283° (decomposes)
- RTECS: BQ4725000
- Powder
Formula:
C₃₇H₃₄Na₂N₂O₉S₃ MW: 792.84 g/mol Melting Pt: 283 °C (dec.) |
MDL Number:
MFCD00012141 CAS Number: 3844-45-9 EINECS: 223-339-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...