Digadolinium trioxide ≥99.9%
Supplier: BeanTown Chemical
Synonyms:
Gadolinium (III) oxide
134210-100G
134210-25G
134210-500G
BT134210-100GEA
159.44
USD
BT134210-100G
BT134210-25G
BT134210-500G
Digadolinium trioxide ≥99.9%
Digadolinium trioxide
CAS: 12064-62-9; EC No: 235-060-9; MDL No: MFCD00011026
- <10 Micron Powder
Formula:
Gd₂O₃ MW: 362.5 g/mol Melting Pt: 2410 Density: 7.407 g/mL |
MDL Number:
MFCD00011026 CAS Number: 12064-62-9 EINECS: 235-060-9 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...