(S)-1-Boc-2-ethylpiperazine ≥97%

Supplier: BeanTown Chemical

Synonyms: (2S)-2-Ethylpiperazine, N1-BOC protected, (S)-tert-Butyl 2-ethylpiperazine-1-carboxylate

134165-1G 134165-250MG
BT134165-1GEA 716 USD
BT134165-1G BT134165-250MG
(S)-1-Boc-2-ethylpiperazine ≥97%
(S)-1-Boc-2-ethylpiperazine

CAS: 325145-35-5; MDL No: MFCD07772100 Solid; Molecular Formula: C11H22N2O2; MW: 214.30


  • Solid

Formula: C₁₁H₂₂N₂O₂
MW: 214.30 g/mol
MDL Number: MFCD07772100
CAS Number: 325145-35-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR