Evans blue ≥85% (dye content)
Supplier: BeanTown Chemical
134155-10G
134155-50G
BT134155-10GEA
55.65
USD
BT134155-10G
BT134155-50G
Evans blue ≥85% (dye content)
Evans blue
CAS: 314-13-6; EC No: 206-242-5; MDL No: MFCD00004021
Powder; Molecular Formula: C34H24N6Na4O14S4; MW: 960.82
- Powder
Formula:
C₃₄H₂₈N₆O₁₄S₄ MW: 960.79 g/mol |
MDL Number:
MFCD00004021 CAS Number: 314-13-6 EINECS: 206-242-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...