2-Amino-3-bromo-5-chloropyridine ≥95%

Supplier: BeanTown Chemical

134110-10G 134110-1G 134110-250MG
BT134110-10GEA 348 USD
BT134110-10G BT134110-1G BT134110-250MG
2-Amino-3-bromo-5-chloropyridine ≥95%
2-Amino-3-bromo-5-chloropyridine

CAS: 26163-03-1; MDL No: MFCD00955627 Solid; Molecular Formula: C5H4BrClN2 ; MW: 207.46


  • Solid

Formula: C₅H₄BrClN₂
MDL Number: MFCD00955627
CAS Number: 26163-03-1

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR