Triiron phosphide ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Iron phosphide

134090-10G 134090-50G
BT134090-10GEA 136 USD
BT134090-10G BT134090-50G
Triiron phosphide ≥99.5% (trace metals basis)
Triiron phosphide

CAS: 12023-53-9; EC No: 234-682-8; MDL No: MFCD00799762 -40 Mesh Powder; Molecular Formula: Fe3P; MW: 198.51


  • -40 Mesh Powder

Formula: Fe₃P
MW: 198.51 g/mol
MDL Number: MFCD00799762
CAS Number: 12023-53-9
EINECS: 234-682-8

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR