Chitin
Supplier: BeanTown Chemical
134085-100G
134085-500G
BT134085-100GEA
51.28
USD
BT134085-100G
BT134085-500G
Chitin
Chitin
CAS: 1398-61-4; EC No: 222-311-2; MDL No: MFCD00161512; RTECS: FM6300000
Powder
- RTECS: FM6300000
- Powder
Formula:
C₃₀H₅₀N₄O₁₉ |
MDL Number:
MFCD00466914 CAS Number: 1398-61-4 EINECS: 222-311-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...