Succinic acid ≥99%

Supplier: BeanTown Chemical

Synonyms: 1,4-Butanedioic acid

134080-10KG 134080-2.5KG 134080-500G
BT134080-10KGEA 477 USD
BT134080-10KG BT134080-2.5KG BT134080-500G
Succinic acid ≥99%
Succinic acid

CAS: 110-15-6; EC No: 203-740-4; MDL No: MFCD00002789; RTECS: WM4900000 Crystalline; Linear Formula: HOOCCH2CH2COOH; Molecular Formula: C4H6O4; MW: 118.09 Melting Point: 184-186°; Boiling Point: 235°; Flash point: 206°C (402°F) Density (g/mL): 1.564


  • RTECS: WM4900000
  • Crystalline

Formula: HOOCCH₂CH₂COOH
MW: 118.09 g/mol
Boiling Pt: 235
Melting Pt: 182
Density: 1.564 g/mL
Flash Pt: 206 °C (402 °F)
MDL Number: MFCD00002789
CAS Number: 110-15-6
EINECS: 203-740-4
UN: 3261
ADR: 8 III
Merck Index: 13,08953
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfd381e6e0e4cbde676a92c9e8d6aacfac635ade&vid=0b14e1b7f82b03468fc82d224a0ee894952eb200&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR