4-Piperidone monohydrate hydrochloride ≥98%

Supplier: BeanTown Chemical

Synonyms: 4,4-Piperidinediol Hydrochloride, 4,4-Dihydroxypiperidine Hydrochloride, 4-Ketopiperidine Monohydrate Hydrochloride, 4-Piperidinone monohydrate hydrochloride, Piperidin-4-one hydrate hydrochloride, 4-Azacyclohexanone Monohydrate Hydrochloride

134075-10G 134075-250G 134075-50G
BT134075-10GEA 43.09 USD
BT134075-10G BT134075-250G BT134075-50G
4-Piperidone monohydrate hydrochloride ≥98%
4-Piperidone monohydrate hydrochloride

CAS: 40064-34-4; EC No: 254-779-9; MDL No: MFCD00012776 Crystalline; Molecular Formula: C5H9NO·HCl·H2O; MW: 153.61 Melting Point: 94-96°


  • Crystalline

Formula: C₅H₉NO·xHCl·yH₂O
MW: 99.13 g/mol (free base)
Melting Pt: 94…96 °C
MDL Number: MFCD00012776
CAS Number: 40064-34-4
EINECS: 254-779-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR