Uridine 5′-monophosphate disodium salt ≥99%

Supplier: BeanTown Chemical

Synonyms: U 5′-P, Disodium uridine-5-monophosphate, Sodium ((2R,3S,4R,5R)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl phosphate, UMP, 5′-UMP-Na2, di-Sodium uridine-5-monophosphate, 5'-UMP 2Na, 5'-Uridylic acid disodium salt, Uridylic acid disodium salt, 5'-UMP disodium salt, Disodium UMP

134070-100G 134070-25G 134070-5G
BT134070-100GEA 821 USD
BT134070-100G BT134070-25G BT134070-5G
Uridine 5′-monophosphate disodium salt ≥99%
Uridine 5′-monophosphate disodium salt

CAS: 3387-36-8; EC No: 222-211-9; MDL No: MFCD00006525; RTECS: YU7975000 Solid; Molecular Formula: C9H11N2O9P; MW: 368.14 Melting Point: 209° (decomposes)


  • RTECS: YU7975000
  • Solid

Formula: C₉H₁₁N₂Na₂O₉P
MW: 368.15 g/mol
Melting Pt: 209 °C (dec.)
MDL Number: MFCD00006525
CAS Number: 3387-36-8
EINECS: 222-211-9

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR