tert-Butyl ethyl malonate ≥95%
Supplier: BeanTown Chemical
134065-100G
134065-25G
134065-5G
BT134065-100GEA
717.97
USD
BT134065-100G
BT134065-25G
BT134065-5G
tert-Butyl ethyl malonate ≥95%
tert-Butyl ethyl malonate
CAS: 32864-38-3; EC No: 251-269-8; MDL No: MFCD00009193
Liquid; Linear Formula: (CH3)3COCOCH2COOCH2CH3; Molecular Formula: C9H16O4; MW: 188.22
Boiling Point: 83-85°/8 mmHg; Flash point: 87°C (189°F)
Density (g/mL): 0.994; Refractive Index: 1.416
- Refractive index: 1.416
- Liquid
Formula:
C₉H₁₆O₄ Boiling Pt: 83…85 °C/8 mmHg Density: 0.994 g/mL Flash Pt: 87 °C (189 °F) |
MDL Number:
MFCD00009193 CAS Number: 32864-38-3 EINECS: 251-269-8 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...