3-Bromoisonicotinonitrile ≥95%

Supplier: BeanTown Chemical

134015-1G 134015-250MG
BT134015-1GEA 523 USD
BT134015-1G BT134015-250MG
3-Bromoisonicotinonitrile ≥95%
3-Bromoisonicotinonitrile

CAS: 13958-98-0; MDL No: MFCD00234143 Solid; Molecular Formula: C6H3BrN2; MW: 183.01


  • Solid

Formula: C₆H₃BrN₂
MW: 183.01 g/mol
MDL Number: MFCD00234143
CAS Number: 13958-98-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR