Praseodymium(III,IV) oxide ≥99.995% (trace rare earth metals basis)
Supplier: BeanTown Chemical
134005-25G
134005-5G
BT134005-25GEA
268
USD
BT134005-25G
BT134005-5G
Praseodymium(III,IV) oxide ≥99.995% (trace rare earth metals basis)
Praseodymium(III,IV) oxide
CAS: 12037-29-5; EC No: 234-857-9; MDL No: MFCD00011178
Powder; Linear Formula: Pr6O11; MW: 1021.43
Melting Point: 2500°
Density (g/mL): 6.5
- Powder
Formula:
O₁₁Pr₆ MW: 1021.44 g/mol Melting Pt: 2500 °C Density: 6.5 g/mL |
MDL Number:
MFCD00011178 CAS Number: 12037-29-5 EINECS: 234-857-9 |
Specification Test Results
Assay | 99.995% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...