Niobium ≥99.5% (trace metals basis), rods Ø 6.35 mm, annealed
Supplier: BeanTown Chemical
133985-100CM
133985-25CM
133985-5CM
BT133985-100CMEA
455
USD
BT133985-100CM
BT133985-25CM
BT133985-5CM
Niobium ≥99.5% (trace metals basis), rods Ø 6.35 mm, annealed
Niobium
CAS: 7440-03-1; EC No: 231-113-5; MDL No: MFCD00011126
Powder; Molecular Formula: Nb ; MW: 92.91
Melting Point: 2468°; Boiling Point: 4742°
Density (g/mL): 8.57
- Powder
Formula:
Nb Boiling Pt: 4742 °C Melting Pt: 2468 °C Density: 8.57 g/mL |
MDL Number:
MFCD00011126 CAS Number: 7440-03-1 EINECS: 231-113-5 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...