2,5-Dibromopyridine ≥97%

Supplier: BeanTown Chemical

133965-10G 133965-50G
BT133965-10GEA 75 USD
BT133965-10G BT133965-50G
2,5-Dibromopyridine ≥97%
2,5-Dibromopyridine

CAS: 624-28-2; EC No: 210-839-6; MDL No: MFCD00006221 Powder; Molecular Formula: C5H3Br2N ; MW: 236.89 Melting Point: 92-95°


  • Powder

Formula: C₅H₃Br₂N
MW: 236.89 g/mol
Melting Pt: 92…95 °C
MDL Number: MFCD00006221
CAS Number: 624-28-2
EINECS: 210-839-6

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR