3-Bromo-1-(trimethylsilyl)-1-propyne ≥98%
Supplier: BeanTown Chemical
Synonyms:
3-(Trimethylsilyl)propargyl bromide, (3-Bromoprop-1-yn-1-yl)trimethylsilane
133940-25G
133940-5G
BT133940-25GEA
307
USD
BT133940-25G
BT133940-5G
3-Bromo-1-(trimethylsilyl)-1-propyne ≥98%
3-Bromo-1-(trimethylsilyl)-1-propyne
CAS: 38002-45-8; MDL No: MFCD00134460
Liquid; Linear Formula: BrCH2C≡CSi(CH3)3; Molecular Formula: C6H11BrSi; MW: 191.14
Boiling Point: 44-45°/2 mmHg; Flash point: 63°C (145°F)
Density (g/mL): 1.17; Refractive Index: 1.483
- Refractive index: 1.483
- Liquid
Formula:
C₆H₁₁BrSi Boiling Pt: 44…45 °C/2 mmHg Density: 1.17 g/mL Flash Pt: 63 °C (145 °F) |
MDL Number:
MFCD00134460 CAS Number: 38002-45-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...