Diindium trisulfate ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Diindium tris(sulphate), Indium(III) sulphate, Indium (III) sulphate
133890-100G
133890-25G
BT133890-100GEA
575
USD
BT133890-100G
BT133890-25G
Diindium trisulfate ≥99.99% (trace metals basis)
Diindium trisulphate
CAS: 13464-82-9; EC No: 236-689-1; MDL No: MFCD00011059
Powder; Linear Formula: In2(SO4)3; MW: 517.83
Hygroscopic
- Hygroscopic
- Powder
Formula:
O₁₂In₂S₃ MW: 517.83 g/mol |
MDL Number:
MFCD00011059 CAS Number: 13464-82-9 EINECS: 236-689-1 |
Specification Test Results
Assay | 99.99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...