2,4,6-Tris(2-pyridyl)-1,3,5-triazine ≥98%

Supplier: BeanTown Chemical

Synonyms: 2,4,6-Tri(2-pyridyl)-s-triazine, 2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 2,4,6-Tris(2-pyridyl)-s-triazine

133855-100G 133855-25G 133855-5G
BT133855-100GEA 1050 USD
BT133855-100G BT133855-25G BT133855-5G
2,4,6-Tris(2-pyridyl)-1,3,5-triazine ≥98%
2,4,6-Tris(2-pyridyl)-1,3,5-triazine

CAS: 3682-35-7; EC No: 222-965-9; MDL No: MFCD00006045; RTECS: XZ2050000 Powder; Molecular Formula: C18H12N6; MW: 312.33 Melting Point: 247-249°


  • RTECS: XZ2050000
  • Powder

Formula: C₁₈H₁₂N₆
MW: 312.33 g/mol
Melting Pt: 247…249 °C
MDL Number: MFCD00006045
CAS Number: 3682-35-7
EINECS: 222-965-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR