Chlorobis(cyclooctene)iridium(I) dimer

Supplier: BeanTown Chemical

Synonyms: Chlorobis(cyclooctene)iridium(I)dimer

133845-1G 133845-5G
BT133845-1GEA 235 USD
BT133845-1G BT133845-5G
Chlorobis(cyclooctene)iridium(I) dimer
Chlorobis(cyclooctene)iridium(I) dimer

CAS: 12246-51-4; MDL No: MFCD00213465 Solid; Molecular Formula: C32H56Cl2Ir2; MW: 896.13 Melting Point: 160-165°


  • Solid

Formula: C₃₂H₅₆Cl₂Ir₂
Melting Pt: 160…165 °C
MDL Number: MFCD00213465
CAS Number: 12246-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR