Silver ≥99.9%, casting grain 1.6-6.35 mm

Supplier: BeanTown Chemical

133835-100G 133835-25G
BT133835-100GEA 459 USD
BT133835-100G BT133835-25G
Silver ≥99.9%, casting grain 1.6-6.35 mm
Silver

CAS: 7440-22-4; EC No: 231-131-3; MDL No: MFCD00003397; RTECS: VW3500000 Solid; Molecular Formula: Ag; MW: 107.87 Melting Point: 960°; Boiling Point: 2212° Density (g/mL): 10.49


  • RTECS: VW3500000
  • Solid

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.49 g/mL
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR