4-Bromo-2-fluorobenzoic acid ≥98%

Supplier: BeanTown Chemical

133820-25G 133820-5G
BT133820-25GEA 507 USD
BT133820-25G BT133820-5G
4-Bromo-2-fluorobenzoic acid ≥98%
4-Bromo-2-fluorobenzoic acid

CAS: 112704-79-7; MDL No: MFCD00143263 Crystalline/Powder; Molecular Formula: C7H4BrFO2; MW: 219.01 Melting Point: 211-215°


  • Crystalline/Powder

Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
Melting Pt: 211…215 °C
MDL Number: MFCD00143263
CAS Number: 112704-79-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR