tert-Butyl methyl malonate ≥95%

Supplier: BeanTown Chemical

Synonyms: tert.-Butyl methyl malonate

133810-10ML 133810-2ML
BT133810-10MLEA 175 USD
BT133810-10ML BT133810-2ML
tert-Butyl methyl malonate ≥95%
tert-Butyl methyl malonate

CAS: 42726-73-8; EC No: 255-919-1; MDL No: MFCD00042856 Liquid; Linear Formula: CH3OCOCH2COOC(CH3)3; Molecular Formula: C8H14O4; MW: 174.19 Melting Point: -10°; Boiling Point: 80°/11 mmHg; Flash point: 79°C (174°F) Density (g/mL): 1.03; Refractive Index: 1.415


  • Refractive index: 1.415
  • Liquid

Formula: C₈H₁₄O₄
MW: 174.19 g/mol
Boiling Pt: 80 °C/11 mmHg
Melting Pt: –10 °C
Density: 1.03 g/mL
Flash Pt: 79 °C (174 °F)
MDL Number: MFCD00042856
CAS Number: 42726-73-8
EINECS: 255-919-1

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR