2-Mercaptobenzoxazole ≥98%
Supplier: BeanTown Chemical
Synonyms:
Benzo[d]oxazole-2-thiol
133800-100G
133800-25G
BT133800-100GEA
111
USD
BT133800-100G
BT133800-25G
2-Mercaptobenzoxazole ≥98%
2-Mercaptobenzoxazole
CAS: 2382-96-9; EC No: 219-191-9; MDL No: MFCD00005769; RTECS: DM4882500
Crystalline; Molecular Formula: C7H5NOS; MW: 151.19
Melting Point: 192-195°
- RTECS: DM4882500
- Crystalline
Formula:
C₇H₅NOS MW: 151.19 g/mol Melting Pt: 192…195 °C |
MDL Number:
MFCD00005769 CAS Number: 2382-96-9 EINECS: 219-191-9 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...