Sodium hypophosphite monohydrate
Supplier: BeanTown Chemical
Synonyms:
Sodium phosphinate monohydrate
133780-1KG
133780-250G
BT133780-1KGEA
92.94
USD
BT133780-1KG
BT133780-250G
Sodium hypophosphite monohydrate
Sodium hypophosphite monohydrate
CAS: 10039-56-2; EC No: 231-669-9; MDL No: MFCD00149207; RTECS: SZ5640000
Powder; Linear Formula: NaH2PO2·H2O; MW: 105.99
Hygroscopic
- RTECS: SZ5640000
- Hygroscopic
- Powder
Formula:
NaH₂PO₂·H₂O MW: 105.99 g/mol |
MDL Number:
MFCD06252379 CAS Number: 10039-56-2 EINECS: 231-669-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...