2-Chloro-4-(trifluoromethyl)phenylboronic acid ≥96%

Supplier: BeanTown Chemical

Synonyms: (2-Chloro-4-(trifluoromethyl)phenyl)metaboric acid, Dihydroxy(2-chloro-4-(trifluoromethyl)phenyl)borane, 2-Chloro-4-(trifluoromethyl)benzeneboronic acid

133775-1G 133775-5G
BT133775-1GEA 102 USD
BT133775-1G BT133775-5G
2-Chloro-4-(trifluoromethyl)phenylboronic acid ≥96%
2-Chloro-4-(trifluoromethyl)phenylboronic acid

CAS: 254993-59-4; MDL No: MFCD02684337 Solid; Molecular Formula: C7H5BClF3O2; MW: 224.38


  • Solid

Formula: C₇H₅BClF₃O₂
MW: 224.37 g/mol
MDL Number: MFCD02684337
CAS Number: 254993-59-4

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR