2,2-Bis(hydroxymethyl)propionic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 3-Hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid

133765-100G 133765-500G
BT133765-100GEA 32 USD
BT133765-100G BT133765-500G
2,2-Bis(hydroxymethyl)propionic acid ≥98%
2,2-Bis(hydroxymethyl)propionic acid

CAS: 4767-03-7; EC No: 225-306-3; MDL No: MFCD00004199 Crystalline; Molecular Formula: C5H10O4; MW: 134.13 Melting Point: 189-191° Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: C₅H₁₀O₄
MW: 134.13 g/mol
Melting Pt: 189…191 °C
MDL Number: MFCD00004199
CAS Number: 4767-03-7
EINECS: 225-306-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR