Ethyl-2-bromooctanoate ≥98%

Supplier: BeanTown Chemical

Synonyms: Ethyl 2-bromooctanoate

133755-100G 133755-25G
BT133755-100GEA 130 USD
BT133755-100G BT133755-25G
Ethyl-2-bromooctanoate ≥98%
Ethyl-2-bromooctanoate

CAS: 5445-29-4; EC No: 226-647-0; MDL No: MFCD00042603 Liquid; Molecular Formula: C10H19BrO2 ; MW: 251.16 Boiling Point: 137-140°/25 mmHg; Flash point: 107°C (225°F) Density (g/mL): 1.167; Refractive Index: 1.452


  • Refractive index: 1.452
  • Liquid

Formula: C₁₀H₁₉BrO₂
MW: 251.164 g/mol
Boiling Pt: 137…140 °C/25 mmHg
Density: 1.167 g/mL
Flash Pt: 107 °C (225 °F)
MDL Number: MFCD00042603
CAS Number: 5445-29-4
EINECS: 226-647-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR