2-Bromo-6-methylpyridine ≥98%
Supplier: BeanTown Chemical
Synonyms:
6-Bromo-2-picoline, 2-Bromo-6-picoline
133735-25G
133735-5G
BT133735-25GEA
290
USD
BT133735-25G
BT133735-5G
2-Bromo-6-methylpyridine ≥98%
2-Bromo-6-methylpyridine
CAS: 5315-25-3; EC No: 226-173-4; MDL No: MFCD00040743
Liquid; Molecular Formula: C6H6BrN; MW: 172.02
Boiling Point: 102-103°/20 mmHg; Flash point: 97°C (207°F)
Density (g/mL): 1.512; Refractive Index: 1.562
- Refractive index: 1.562
- Liquid
Formula:
C₆H₆BrN MW: 172.02 g/mol Boiling Pt: 102…103 °C/20 mmHg Density: 1.512 g/mL Flash Pt: 97 °C (207 °F) |
MDL Number:
MFCD00040743 CAS Number: 5315-25-3 EINECS: 226-173-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...