Tween® 60 (Polysorbate)
Supplier: BeanTown Chemical
133675-100ML
133675-500ML
BT133675-100MLEA
27.54
USD
BT133675-100ML
BT133675-500ML
Tween® 60 (Polysorbate)
Tween® 60 (Polysorbate)
CAS: 9005-67-8; MDL No: MFCD01780110; RTECS: WG2934000
Liquid
Boiling Point:
- RTECS: WG2934000
- Refractive index: 1.474
- Liquid
Formula:
C₆₄H₁₂₆O₂₆ Boiling Pt: >100 °C Density: 1.044 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD01780110 CAS Number: 9005-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...