L(+)-Lysine monohydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: L-(+)-Lysine, Hydrate, (S)-(+)-2,6-Diaminohexanoic acid monohydrate, (S)-(+)-Lysine monohydrate, L-Lysine hydrate, (S)-(+)-2,6-Diaminocaproic acid monohydrate, H-Lys-OH monohydrate, K monohydrate, H-Lys-OH·H₂O, Lys monohydrate

133655-100G 133655-25G 133655-500G
BT133655-100GEA 157.56 USD
BT133655-100G BT133655-25G BT133655-500G
L(+)-Lysine monohydrate ≥98%
L(+)-Lysine monohydrate

CAS: 39665-12-8; EC No: 200-294-2; MDL No: MFCD00151035; RTECS: OL5540000 Powder; Molecular Formula: C6H14N2O2·H2O; MW: 164.20 Optical Rotation: [α]20/D +22.5±1°, c = 2% in 6M HCl


  • RTECS: OL5540000
  • Optical rotation: [α]20/D +22.5±1°, c = 2% in 6M HCl
  • Powder

Formula: H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O
MW: 164.2 g/mol
MDL Number: MFCD00151035
CAS Number: 39665-12-8
EINECS: 200-294-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR