(S)-(-)-α,α-Diphenylprolinol ≥98%
Supplier: BeanTown Chemical
133645-1G
133645-5G
BT133645-1GEA
81
USD
BT133645-1G
BT133645-5G
(S)-(-)-α,α-Diphenylprolinol ≥98%
(S)-(-)-α,α-Diphenylprolinol
CAS: 112068-01-6; MDL No: MFCD00075506
Solid; Molecular Formula: C17H19NO ; MW: 253.34
Melting Point: 77-80°
Optical Rotation: [α]20/D -67°, c = 3 in chloroform
- Optical rotation: [α]20/D -67°, c = 3 in chloroform
- Solid
Formula:
C₁₇H₁₉NO Melting Pt: 77…80 °C |
MDL Number:
MFCD00075506 CAS Number: 112068-01-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...