4-Iodophenol ≥98%
Supplier: BeanTown Chemical
133640-100G
133640-25G
133640-5G
BT133640-100GEA
263
USD
BT133640-100G
BT133640-25G
BT133640-5G
4-Iodophenol ≥98%
4-Iodophenol
CAS: 540-38-5; EC No: 208-745-5; MDL No: MFCD00002327; RTECS: SL5600000
Powder; Molecular Formula: C6H5IO; MW: 220.01
Melting Point: 92-94°; Boiling Point: 138°/5 mmHg
Light Sensitive
- RTECS: SL5600000
- Light Sensitive
- Powder
Formula:
C₆H₅IO Boiling Pt: 138 °C/5 mmHg Melting Pt: 92…94 °C |
MDL Number:
MFCD00002327 CAS Number: 540-38-5 EINECS: 208-745-5 UN: 3261 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...