Benserazide hydrochloride ≥98%

Supplier: BeanTown Chemical

Synonyms: Benserazide HCI, DL-Serine 2-(2,3,4-Trihydroxybenzyl)hydrazide Hydrochloride, 2-Amino-3-hydroxy-N'-(2,3,4-trihydroxybenzyl)propanehydrazide hydrochloride

133590-1G 133590-5G
BT133590-1GEA 81 USD
BT133590-1G BT133590-5G
Benserazide hydrochloride ≥98%
Benserazide hydrochloride

CAS: 14919-77-8; EC No: 238-991-9; MDL No: MFCD00078571; RTECS: VT9632300 Powder; Molecular Formula: C10H15N3O5·HCl; MW: 293.70 Light Sensitive


  • RTECS: VT9632300
  • Light Sensitive
  • Powder

Formula: C₁₀H₁₅N₃O₅
MW: 293.70 g/mol
MDL Number: MFCD00078571
CAS Number: 14919-77-8
EINECS: 238-991-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR