Mucic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
Galactaric acid, rel-(2R,3S,4R,5S)-2,3,4,5-Tetrahydroxyhexanedioic acid, (2R,3S,4R,5S)-2,3,4,5-Tetrahydroxyhexanedioic acid
133565-100G
133565-500G
BT133565-100GEA
50
USD
BT133565-100G
BT133565-500G
Mucic acid ≥98%
Mucic acid
CAS: 526-99-8; EC No: 208-404-0; MDL No: MFCD00004239; RTECS: LW5180000
Powder; Linear Formula: HOOC(CHOH)4COOH; Molecular Formula: C6H10O8; MW: 210.14
Melting Point: 220-225° (decomposes)
- RTECS: LW5180000
- Powder
Formula:
C₆H₁₀O₈ MW: 210.14 g/mol Melting Pt: 220…225 °C (dec.) |
MDL Number:
MFCD00004239 CAS Number: 526-99-8 EINECS: 208-404-0 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...