3,5-Dibromobenzaldehyde ≥98%

Supplier: BeanTown Chemical

133555-1G 133555-25G 133555-5G
BT133555-1GEA 31 USD
BT133555-1G BT133555-25G BT133555-5G
3,5-Dibromobenzaldehyde ≥98%
3,5-Dibromobenzaldehyde

CAS: 56990-02-4; MDL No: MFCD00156887 Powder; Molecular Formula: C7H4Br2O; MW: 263.91 Melting Point: 84-88° Air Sensitive


  • Air Sensitive
  • Powder

Formula: C₇H₄Br₂O
MW: 263.92
Melting Pt: 84…88 °C
MDL Number: MFCD00156887
CAS Number: 56990-02-4

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR