3-(Trimethoxysilyl)propyl methacrylate ≥97%
Supplier: BeanTown Chemical
Synonyms:
γ-methacryloxypropyl-trimethoxysilane, 3-(Methacryloyloxy)propyltrimethoxysilane
133540-100G
133540-500G
BT133540-100GEA
37.55
USD
BT133540-100G
BT133540-500G
3-(Trimethoxysilyl)propyl methacrylate ≥97%
3-(Trimethoxysilyl)propyl methacrylate
CAS: 2530-85-0; EC No: 219-785-8; MDL No: MFCD00008593; RTECS: UC0230000
Liquid; Molecular Formula: C10H20O5Si; MW: 248.35
Boiling Point: 190°; Flash point: 92°C (198°F)
Density (g/mL): 1.045; Refractive Index: 1.431
Moisture Sensitive
- RTECS: UC0230000
- Refractive index: 1.431
- Moisture Sensitive
- Liquid
Formula:
C₁₀H₂₀O₅Si MW: 248.35 g/mol Boiling Pt: 190 °C Density: 1.045 g/mL Flash Pt: 92 °C (198 °F) |
MDL Number:
MFCD00008593 CAS Number: 2530-85-0 EINECS: 219-785-8 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...